missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cleaning Solution, Chromic-Sulfuric Acid Solution, Ricca Chemical
For Cleaning Laboratory Glassware
Supplier: Ricca Chemical Company 215016
Description
- Clear, brown liquid
- 7789-12-0 (Sodium Dichromate Dihydrate); 1333-82-0 (Chromium (VI) Trioxide); 7664-93-9 (Sulfuric Acid)
Chemical Identifiers
| Cleaning Solution | |
| Liquid | |
| Non-sterile | |
| ∼300°C | |
| Chromium (VI) Trioxide 0.8 to 1%, Sulfuric Acid 97 to 99 %, Water, Deionized 1 to 1.2%, Sodium Dichr |
| No | |
| Unscented | |
| Dark Brown | |
| OSHA, 29 CFR 1910.1200, TSCA | |
| 15°C to 30°C |
Specifications
| Cleaning Solution | |
| Liquid | |
| Non-sterile | |
| ∼300°C | |
| Chromium (VI) Trioxide 0.8 to 1%, Sulfuric Acid 97 to 99 %, Water, Deionized 1 to 1.2%, Sodium Dichr | |
| 500 mL |
| No | |
| Unscented | |
| Dark Brown | |
| OSHA, 29 CFR 1910.1200, TSCA | |
| 15°C to 30°C |