Learn More
Promega Riboprobe™ Systems
The Riboprobe Systems are designed to prepare high-specific-activity single-stranded RNA probes or microgram quantities of defined RNA transcripts from cloned DNA inserts.
Supplier: Promega P1440
Includes: RNA polymerase(s); pGEM™ Express Positive Control Template; RQ1 RNase-free DNase; Recombinant RNasin™ Ribonuclease Inhibitor; rATP; rCTP; rGTP; rUTP; DTT; Transcription-optimized buffer, 5X; Nuclease-free water
Description
- Produce High-Specific-Activity RNA Probes or Defined RNA Transcripts
- Systems available with SP6 RNA Polymerase, T7 RNA Polymerase or T3 RNA Polymerase
- SP6, T7 and T3 RNA Polymerases are extremely promoter-specific, allowing production of virtually homogeneous RNA using plasmid DNA as a template
- Allow synthesis of RNA corresponding to the coding or noncoding strand of cloned DNA from a single plasmid construct
- U.S. Patent 5,552,302; Australian Patent 646803 and other patents.
- Australian Patents 616881 and 641261 and other pending and issued patents, which claim vectors encoding a portion of human placental ribonuclease inhibitor, are exclusively licensed to Promega Corporation.
Specifications
| Store at -20°C. | |
| 500U RNA Polymerase (T7), 5μg pGEM™ express positive control template, 500μL transcription optimized 5X Buffer, 2500U recombinant RNasin™ RNase inhibitor, 100mM 100 | |
| 1 System |
| Generation of high-specific-activity RNA probes, generation of microgram quantities of RNA transcripts, generation of RNA processing substrates, generation of antisense RNA | |
| Riboprobe System—T7 |