Learn More
Worthington Biochemical RIBONUCLEASE T1 CHROMA 100KU
Catalog No. 50592457
Encompass
Highly purified, microbial (non-mammalian) RNase prepared with non-animal components. Supplied as a solution in 2.8 M ammonium sulfate. 2603-05-7
Catalog No. 50592457 Supplier Worthington Biochemical Supplier No. LS01485
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Highly purified, microbial (non-mammalian) RNase prepared with non-animal components. Supplied as a solution in 2.8 M ammonium sulfate. 2603-05-7