missing translation for 'onlineSavingsMsg'
Learn More

Riboflavin, USP, 98-102%, Spectrum™ Chemical

Catalog No. 18606094
Change view
Click to view available options
Quantity:
500 g
1 kg
12 kg
25 kg
50 kg
Packaging:
Amber Glass Bottle
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18606094 50 kg Amber Glass Bottle
18606090 500 g Amber Glass Bottle
18606091 1 kg Amber Glass Bottle
18606092 12 kg Amber Glass Bottle
18606093 25 kg Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 18606094 Supplier Spectrum Chemical Mfg Cor Supplier No. RI10350KGBL
Get Up to $500 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

RI103, 83-88-5, C17H20N4O6

Spectrum™ Chemical Riboflavin, USP is also referred to as vitamin B2 and used as a dietary supplement. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 83-88-5
Molecular Formula C17H20N4O6
Molecular Weight (g/mol) 376.37
MDL Number MFCD00005022
InChI Key AUNGANRZJHBGPY-QTZZOOGMNA-N
IUPAC Name 7,8-dimethyl-10-[(2S,3S,4R)-2,3,4,5-tetrahydroxypentyl]-2H,3H,4H,10H-benzo[g]pteridine-2,4-dione
SMILES CC1=C(C)C=C2N(C[C@H](O)[C@H](O)[C@H](O)CO)C3=NC(=O)NC(=O)C3=NC2=C1

Specifications

CAS Max % 1
Ignition Residue 0.003
Loss on Drying 0.015
Packaging Amber Glass Bottle
Quantity 50 kg
Specific Rotation -115° to -135°
Percent Purity 98 to 102%
Grade USP
Physical Form Crystalline Powder or Lumps
Chemical Name or Material Riboflavin