missing translation for 'onlineSavingsMsg'
Learn More

Riboflavin, TCI America™

Catalog No. p-7135664
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
R002025G 25 g
R0020500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. R002025G Supplier TCI America Supplier No. R002025G

Chemical Identifiers

CAS 83-88-5
Molecular Formula C17H20N4O6
Molecular Weight (g/mol) 376.37
MDL Number MFCD00005022
InChI Key AUNGANRZJHBGPY-QTZZOOGMNA-N
Synonym riboflavin, vitamin b2, lactoflavin, riboflavine, vitamin g, lactoflavine, --riboflavin, beflavin, beflavine, flavaxin
PubChem CID 71310809
IUPAC Name 7,8-dimethyl-10-[(2S,3S,4R)-2,3,4,5-tetrahydroxypentyl]-2H,3H,4H,10H-benzo[g]pteridine-2,4-dione
SMILES CC1=C(C)C=C2N(C[C@H](O)[C@H](O)[C@H](O)CO)C3=NC(=O)NC(=O)C3=NC2=C1

Specifications

Quantity 25 g
Formula Weight 376.37
Physical Form Crystalline Powder
Chemical Name or Material Riboflavin
EINECSNumber (5)-3909&(9)-2282
RTECSNumber VJ1400000
TSCA Yes