missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Riboflavin Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™
Description
Cleaning Products, Cosmetics, Food and Beverages, Personal Care and Pharmaceutical (small molecule)
Specifications
Specifications
| CAS | 83-88-5 |
| Physical Form | Neat |
| Linear Formula | C17H20N4O6 |
| MDL Number | MFCD00005022 |
| Melting Point | 290°C |
| Synonym | (-)-Riboflavin; Lactoflavin; Vitamin B2; Vitamin G |
| Type | Certified Reference Material |
| Quantity | 1 g |
| Formula Weight | 376.36 |
| Grade | Certified Reference Material |