missing translation for 'onlineSavingsMsg'
Learn More

Riboflavin, FCC, 98-102%, Spectrum™ Chemical

Catalog No. p-7158137
Change view
Click to view available options
Quantity:
25 g
500 g
1 kg
25 kg
50 kg
Packaging:
Amber Glass Bottle
Fiber Drum
Poly Pail
Catalog No. Quantity Packaging
18606082 25 g Amber Glass Bottle
18606084 500 g Amber Glass Bottle
18606085 1 kg Poly Pail
18606086 25 kg Fiber Drum
18606087 50 kg Fiber Drum
5 options

Catalog No. 18606082

Supplier: Spectrum Chemical Mfg Cor R103225GMGL

Only null left

R1032, 83-88-5, C17H20N4O6

Spectrum™ Chemical Riboflavin, FCC is also referred to as vitamin B2 and used as a dietary supplement. The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 83-88-5
Molecular Formula C17H20N4O6
Molecular Weight (g/mol) 376.37
MDL Number MFCD00005022
InChI Key AUNGANRZJHBGPY-QTZZOOGMNA-N
IUPAC Name 7,8-dimethyl-10-[(2S,3S,4R)-2,3,4,5-tetrahydroxypentyl]-2H,3H,4H,10H-benzo[g]pteridine-2,4-dione
SMILES CC1=C(C)C=C2N(C[C@H](O)[C@H](O)[C@H](O)CO)C3=NC(=O)NC(=O)C3=NC2=C1

Specifications

CAS Max % 1
Ignition Residue 0.003
Loss on Drying 0.015
Packaging Amber Glass Bottle
Quantity 25 g
Specific Rotation +56.5° to +59.5°
Percent Purity 98 to 102%
Grade FCC