missing translation for 'onlineSavingsMsg'
Learn More

Riboflavin, 98%

Catalog No. AAA1176414
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1176414 25 g
AAA1176422 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1176414 Supplier Thermo Scientific Chemicals Supplier No. A1176414
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 83-88-5 | C17H20N4O6 | 376.37 g/mol

Photoinitiator of polymerization for PAGE gelsRiboflavin is used in the preparation of flavin mononucleotide (FMN) and flavin adenine dinucleotide (FAD). It also provides protection against oxidative damage. It is also used as a food coloring and a vitamin. Further, it is employed as a medication, animal feed supplement and nutritional supplement. In addition to this, it is used as a flavoring agent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Photoinitiator of polymerization for PAGE gelsRiboflavin is used in the preparation of flavin mononucleotide (FMN) and flavin adenine dinucleotide (FAD). It also provides protection against oxidative damage. It is also used as a food coloring and a vitamin. Further, it is employed as a medication, animal feed supplement and nutritional supplement. In addition to this, it is used as a flavoring agent.

Solubility
Soluble in water, absolute ethanol and dimethyl sulfoxide. Insoluble in acetone and chloroform.

Notes
Store in a cool place. Incompatible with Strong oxidizing agents, reducing agents, bases, calcium and metallic salts.

Chemical Identifiers

CAS 83-88-5
Molecular Formula C17H20N4O6
Molecular Weight (g/mol) 376.37
MDL Number MFCD00005022
InChI Key AUNGANRZJHBGPY-QTZZOOGMNA-N
Synonym riboflavin, vitamin b2, lactoflavin, riboflavine, vitamin g, lactoflavine, --riboflavin, beflavin, beflavine, flavaxin
PubChem CID 71310809
IUPAC Name 7,8-dimethyl-10-[(2S,3S,4R)-2,3,4,5-tetrahydroxypentyl]benzo[g]pteridine-2,4-dione
SMILES CC1=C(C)C=C2N(C[C@H](O)[C@H](O)[C@H](O)CO)C3=NC(=O)NC(=O)C3=NC2=C1

Specifications

Melting Point 290°C
Odor Odorless
Assay Percent Range 98%
Quantity 25 g
Sensitivity Light sensitive
Merck Index 14,8200
Solubility Information Soluble in water,absolute ethanol and dimethyl sulfoxide. Insoluble in acetone and chloroform.
Formula Weight 376.37
Percent Purity 98%
Chemical Name or Material Riboflavin
EINECSNumber 201-507-1
RTECSNumber VJ1400000
TSCA Yes
Recommended Storage Keep cold

RUO – Research Use Only