missing translation for 'onlineSavingsMsg'
Learn More

United Scientific™ RIA Vial Caps

Catalog No. S15698
Change view
Click to view available options
Color:
Transparent
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
S15698 Transparent
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S15698 Supplier United Scientific™ Supplier No. P10304

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Caps and tubes sold separately and are used in radionimmunoassay.

  • RIA vials are generally used in radioimmunoassay, a scientific method used to test antigens without the need to use a bioassay.
  • The polypropylene vials are translucent and can withstand 3000 RCF during centrifugation.
  • Can be used with common acids, alkalies, and solvents at room temperature. Autoclavable.
  • The polystyrene vials are transparent and can withstand 1400 RCF during centrifugation.
  • Can be used with mild bases and weak acids, but not organic solvents.
  • Non-autoclavable.
  • Caps are sold separately and are compatible with both versions of vials.

Specifications

Color Transparent
Diameter (Metric) Outer 12 mm
Material HDPE
Pre-slit No
Sterility Non-sterile
No. per Pack 500/Pk.
Quantity 500/Cs.
For Use With (Equipment) RIA Vials
Autoclavable Not Autoclavable
Product Type Screw Cap