Learn More
Eisco™ Rhyolite Specimen
Rhyolite Specimen is aphanitic felsic rock, formed during eruptions of thick, viscous magma.
Supplier: Eisco™ ESNG0049PK12
Description
- For use in density comparison with other igneous rocks
- Correlates well to Next Generation Science Standards Evidence Statements
- High silica content makes it difficult for this rock to erupt, leading to explosive eruptions that emit ash and dust into atmosphere
- Evolved chemistry makes it lightweight relative to other igneous rocks, good identifier during classification exercises
Identifiants chimiques
| Igneous Rock, Rhyolite |
Spécifications
| Igneous Rock, Rhyolite | |
| Clear labeled and barcoded bag | |
| 2 to 3 cm |
| Density comparison with other igneous rocks | |
| 12/Pk. |