Learn More
Rhodium powder, -325 mesh, 99.95% (metals basis)
CAS: 7440-16-6 | Rh | 102.91 g/mol
$2282.53 - $2282.53
Chemical Identifiers
| CAS | 7440-16-6 |
|---|---|
| Molecular Formula | Rh |
| Molecular Weight (g/mol) | 102.91 |
| MDL Number | MFCD00011201 MFCD03458392 |
| InChI Key | MHOVAHRLVXNVSD-UHFFFAOYSA-N |
| Synonym | fume, on carbon, rhodium, elemental, rodio, black, rhodium, metal, unii-dmk383dsac, on alumina, rh/al2o3, dmk383dsac |
| PubChem CID | 23948 |
| ChEBI | CHEBI:33359 |
| IUPAC Name | rhodium |
| SMILES | [Rh] |
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 7440-16-6 | |
| 102.91 | |
| MHOVAHRLVXNVSD-UHFFFAOYSA-N | |
| 23948 | |
| rhodium |
| Rh | |
| MFCD00011201 MFCD03458392 | |
| fume, on carbon, rhodium, elemental, rodio, black, rhodium, metal, unii-dmk383dsac, on alumina, rh/al2o3, dmk383dsac | |
| CHEBI:33359 | |
| [Rh] |
Specifications
| 7440-16-6 | |
| 0.5 g | |
| MFCD00011201 MFCD03458392 | |
| fume, on carbon, rhodium, elemental, rodio, black, rhodium, metal, unii-dmk383dsac, on alumina, rh/al2o3, dmk383dsac | |
| [Rh] | |
| 102.91 | |
| CHEBI:33359 | |
| Odorless | |
| (metals basis) |
| Powder | |
| Rh | |
| UN3089 | |
| MHOVAHRLVXNVSD-UHFFFAOYSA-N | |
| rhodium | |
| 23948 | |
| 99.95% | |
| Rhodium powder |
Safety and Handling
GHS H Statement
H228
Flammable solid.
GHS P Statement
P210-P280-P240-P241-P370+P378a
Keep away from heat/sparks/open flames/hot surfaces.
No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
Ground/bond container and receiving equipment.
Use explosion-proof electrical/ventilating/lighting/ equipment.
In case of fire: Use for extinction: CO2, powder or water spray.
H500
DOTInformation : Transport Hazard Class: 4.1; Packing Group: III; Proper Shipping Name: METAL POWDERS, FLAMMABLE, N.O.S.
EINECSNumber : 231-125-0
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only