missing translation for 'onlineSavingsMsg'
Learn More

Rhodium(III) chloride, anhydrous, 48.6-49.2% Rh

CAS: 10049-07-7 | Cl3Rh | 209.26 g/mol

$1408.18 - $6125.16

Chemical Identifiers

CAS 10049-07-7
Molecular Formula Cl3Rh
Molecular Weight (g/mol) 209.26
MDL Number MFCD00011204
InChI Key SONJTKJMTWTJCT-UHFFFAOYSA-K
IUPAC Name rhodium(3+) trichloride
SMILES [Cl-].[Cl-].[Cl-].[Rh+3]
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC419105000
SDS View Documents
Thermo Scientific Chemicals
419105000
500 mg Glass bottle
Each for $1,408.18
Only null left
 
AC419100025
SDS View Documents
Thermo Scientific Chemicals
419100025
2.5 g Glass bottle
Each for $6,125.16
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Reduction reagent
Specifications

Chemical Identifiers

10049-07-7
209.26
SONJTKJMTWTJCT-UHFFFAOYSA-K
[Cl-].[Cl-].[Cl-].[Rh+3]
Cl3Rh
MFCD00011204
rhodium(3+) trichloride

Specifications

717.0°C (730.0 mmHg)
10049-07-7
Crystalline Powder
Cl3Rh
MFCD00011204
15, 8313
SONJTKJMTWTJCT-UHFFFAOYSA-K
rhodium(3+) trichloride
209.26
Glass bottle
5.3800g/mL
450.0°C
Black
500 mg
RhCl3
02,357; 03,242; 06,504; 07,313; 08,435; 11,460; 14,271; 16,292; 17,366
Solubility in water: insoluble. Other solubilities: soluble in alkaline solution
[Cl-].[Cl-].[Cl-].[Rh+3]
209.26
48.6 to 49.2% Rh
5.38
Rhodium(III) chloride, 48.6-49.2% Rh
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes serious eye damage.
Harmful if swallowed.
Suspected of causing genetic defects.
May be corrosive to metals.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Immediately

GHS Signal Word: Danger

EINECSNumber : 233-165-4

RTECSNumber : VI9275000

TSCA : TSCA

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only