missing translation for 'onlineSavingsMsg'
Learn More

Rhodium(II) trifluoroacetate dimer

CAS: 31126-95-1 | C8F12O8Rh2 | 657.88 g/mol

$229.66 - $869.72

Chemical Identifiers

CAS 31126-95-1
Molecular Formula C8F12O8Rh2
Molecular Weight (g/mol) 657.88
MDL Number MFCD00209611
InChI Key SZQVMUPTZFMHQT-UHFFFAOYSA-N
Synonym rhodium ii trifluoroacetate dimer, trifluoroacetic acid rhodium ii salt dimer, dirhodium ii tetra trifluoroacetate, rhodium ii trifluoroacetate dimer, ?rh2 cf3coo 4, bis 2,2,2-trifluoroacetyl oxy rhodio 2,2,2-trifluoroacetate
PubChem CID 10985187
IUPAC Name rhodium;2,2,2-trifluoroacetic acid
SMILES C(=O)(C(F)(F)F)O.C(=O)(C(F)(F)F)O.C(=O)(C(F)(F)F)O.C(=O)(C(F)(F)F)O.[Rh].[Rh]
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC389702500
SDS View Documents
Thermo Scientific Chemicals
389702500
250 mg Glass bottle N/A
Only null left
 
AC389700010
SDS View Documents
Thermo Scientific Chemicals
389700010
1 g Glass bottle N/A
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

31126-95-1
657.88
SZQVMUPTZFMHQT-UHFFFAOYSA-N
10985187
C(=O)(C(F)(F)F)O.C(=O)(C(F)(F)F)O.C(=O)(C(F)(F)F)O.C(=O)(C(F)(F)F)O.[Rh].[Rh]
C8F12O8Rh2
MFCD00209611
rhodium ii trifluoroacetate dimer, trifluoroacetic acid rhodium ii salt dimer, dirhodium ii tetra trifluoroacetate, rhodium ii trifluoroacetate dimer, ?rh2 cf3coo 4, bis 2,2,2-trifluoroacetyl oxy rhodio 2,2,2-trifluoroacetate
rhodium;2,2,2-trifluoroacetic acid

Specifications

Green
Crystalline Powder
C8F12O8Rh2
rhodium ii trifluoroacetate dimer, trifluoroacetic acid rhodium ii salt dimer, dirhodium ii tetra trifluoroacetate, rhodium ii trifluoroacetate dimer, ?rh2 cf3coo 4, bis 2,2,2-trifluoroacetyl oxy rhodio 2,2,2-trifluoroacetate
C(=O)(C(F)(F)F)O.C(=O)(C(F)(F)F)O.C(=O)(C(F)(F)F)O.C(=O)(C(F)(F)F)O.[Rh].[Rh]
657.88
657.88
Glass bottle
31126-95-1
250 mg
MFCD00209611
SZQVMUPTZFMHQT-UHFFFAOYSA-N
rhodium;2,2,2-trifluoroacetic acid
10985187
Authentic
Rhodium(II) trifluoroacetate dimer
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

SDS
Documents

Documents

RUO – Research Use Only