missing translation for 'onlineSavingsMsg'
Learn More

Rhodium(II) acetate dimer, dihydrate, ca 43% Rh

Catalog No. p-41399
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC197612500 250 mg Glass bottle
AC197610010 1 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC197612500 Supplier Thermo Scientific Chemicals Supplier No. 197612500

CAS: 29998-99-0 | C8H16O10Rh2 | 478.02 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 29998-99-0
Molecular Formula C8H16O10Rh2
Molecular Weight (g/mol) 478.02
MDL Number MFCD00149212
Synonym Tetrakis[micron-(acetato-O:O')]diaquadirhodium

Specifications

Color Green
Physical Form Fine Powder
Quantity 250 mg
Linear Formula [(CH3CO2)2RhH2O]2
Fieser 08,434; 13,266; 15,278; 16,289; 17,298; 05,571
Solubility Information Solubility in water: insoluble. Other solubilities: moderately soluble in many polar solvents like eth, anol, acetone, acetic acid, acetonitrile, dmso, ni, tromethane, thf, triethyl phosphate.
Formula Weight 478.02
Percent Purity ∼43% (Rh)
Infrared Spectrum Authentic
Packaging Glass bottle
Chemical Name or Material Rhodium(II) acetate dimer, dihydrate, ca 43% Rh
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement GHS P Statement:
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

RUO – Research Use Only