missing translation for 'onlineSavingsMsg'
Learn More

Rhodium on activated carbon, 5% Rh, unreduced, dry

Catalog No. p-41381
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC195440010 1 g Glass bottle
AC195440050 5 g Glass bottle
AC195440250 25 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC195440010 Supplier Thermo Scientific Chemicals Supplier No. 195440010
Only null left

CAS: 7440-16-6 | Rh | 102.91 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7440-16-6 , 7440-44-0
Molecular Formula Rh
Molecular Weight (g/mol) 102.91
MDL Number MFCD00011201 MFCD03458392
InChI Key MHOVAHRLVXNVSD-UHFFFAOYSA-N
Synonym fume, on carbon, rhodium, elemental, rodio, black, rhodium, metal, unii-dmk383dsac, on alumina, rh/al2o3, dmk383dsac
PubChem CID 23948
ChEBI CHEBI:33359
IUPAC Name rhodium
SMILES [Rh]

Specifications

Color Black to Black-Gray
Quantity 1 g
Packaging Glass bottle
Particle Size Typical particle size distribution: D50: 20 Micron, <8 Micron: 17%, <16 Micron: 39%,< 32 Micron: 67%, <64 Micron: 90%, <128 Micron: 99%
Form Powder
Fieser 01,982; 03,326; 04,418; 06,503; 08,433
Merck Index 15, 8310
Formula Weight 102.91
Percent Purity 5% (Rh)
Chemical Name or Material Rhodium on activated carbon, 5% Rh
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Flammable solid.
Precautionary Statement GHS P Statement:
Keep container tightly closed.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

RUO – Research Use Only