missing translation for 'onlineSavingsMsg'
Learn More

Rhodanine, Reagent Grade, LabChem™

Catalog No. LC222408
Change view
Click to view available options
Quantity:
25 g
Packaging:
Amber Glass
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
LC222408 25 g Amber Glass
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. LC222408 Supplier LabChem Supplier No. LC222408

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 141-84-4
Molecular Formula C3H3NOS2
Molecular Weight (g/mol) 133.183
InChI Key KIWUVOGUEXMXSV-UHFFFAOYSA-N
Synonym rhodanine, 2-thioxothiazolidin-4-one, 2-thioxo-4-thiazolidinone, rhodaninic acid, 4-thiazolidinone, 2-thioxo, 2-thioxo-1,3-thiazolidin-4-one, rhodanic acid, rhodanin, rodanin, 4-oxo-2-thioxothiazoline
PubChem CID 1201546
ChEBI CHEBI:8830
IUPAC Name 2-sulfanylidene-1,3-thiazolidin-4-one
SMILES C1C(=O)NC(=S)S1

Specifications

CAS Max % 100
Color Yellow
Density 0.868g/cm3
Packaging Amber Glass
Linear Formula C3H3NOS2
Quantity 25 g
Solubility Information Soluble in water
Formula Weight 133.19
Grade Reagent
Physical Form Powder
Chemical Name or Material Rhodanine
Show More Show Less
Hazard Category Danger
Hazard Statement GHS H Statement
Harmful if swallowed.
Causes serious eye damage.
Precautionary Statement GHS P Statement
Wear protective gloves, eye protection.
Wash exposed skin thoroughly after handling.
Do not eat, drink or smoke when using this product.
f swallowed: Rinse mouth.
Call a poison center/doctor if you feel unwell.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Immediately call a poison center/doctor.
Dispose of contents/container to comply with local, state and federal regulations.
EINECSNumber 205-505-1
Recommended Storage Room Temperature