missing translation for 'onlineSavingsMsg'
Learn More

Rhodanine, 99%

Catalog No. p-190976
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC152600250 25 g Glass bottle
AC152601000 100 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC152600250 Supplier Thermo Scientific Chemicals Supplier No. 152600250
Only null left

CAS: 141-84-4 | C3H3NOS2 | 133.183 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 141-84-4
Molecular Formula C3H3NOS2
Molecular Weight (g/mol) 133.183
MDL Number MFCD00005488
InChI Key KIWUVOGUEXMXSV-UHFFFAOYSA-N
Synonym 2-Thioxo-4-thiazolidinone
PubChem CID 1201546
ChEBI CHEBI:8830
IUPAC Name 2-sulfanylidene-1,3-thiazolidin-4-one
SMILES C1C(=O)NC(=S)S1

Specifications

Melting Point 167°C to 170°C
Assay Percent Range 99%
Beilstein 27, 242
Merck Index 15, 8308
Infrared Spectrum Authentic
Formula Weight 133.2
Percent Purity ≥98.5%
Packaging Glass bottle
Color Beige to Yellow
Quantity 25 g
Physical Form Fine Crystalline Powder
Chemical Name or Material Rhodanine, 99%
Show More Show Less
Hazard Statement GHS H Statement:
Harmful if swallowed.
Causes serious eye irritation.
Precautionary Statement GHS P Statement:
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 205-505-1
RTECSNumber VI7700000
TSCA TSCA

RUO – Research Use Only