Learn More
Rhodanine, 98+%
CAS: 141-84-4 | C3H3NOS2 | 133.183 g/mol
Supplier: Thermo Scientific Chemicals A1829322
Description
Rhodanine inhibits the multiplication of echovirus 12 and the development of virus-induced morphologic changes. It has been used in tannase assay in cultures of tannin degrading fungi. It possess anticonvulsant, antibacterial, antiviral and antidiabetic activities.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsRhodanine inhibits the multiplication of echovirus 12 and the development of virus-induced morphologic changes. It has been used in tannase assay in cultures of tannin degrading fungi. It possess anticonvulsant, antibacterial, antiviral and antidiabetic activities.
Solubility
Soluble in water (2.25 g/L at 20°C).
Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
Specifications
| Rhodanine | |
| 166°C to 171°C | |
| Odorless | |
| MFCD00005488 | |
| 110701 | |
| rhodanine, 2-thioxothiazolidin-4-one, 2-thioxo-4-thiazolidinone, rhodaninic acid, 4-thiazolidinone, 2-thioxo, 2-thioxo-1,3-thiazolidin-4-one, rhodanic acid, rhodanin, rodanin, 4-oxo-2-thioxothiazoline | |
| KIWUVOGUEXMXSV-UHFFFAOYSA-N | |
| 2-sulfanylidene-1,3-thiazolidin-4-one | |
| 1201546 | |
| 133.19 |
| 141-84-4 | |
| 0.868 | |
| C3H3NOS2 | |
| 100 g | |
| 14,8184 | |
| Soluble in water (2.25g/L at 20°C). | |
| C1C(=O)NC(=S)S1 | |
| 133.183 | |
| CHEBI:8830 | |
| ≥98% |
Chemical Identifiers
| 141-84-4 | |
| 133.183 | |
| KIWUVOGUEXMXSV-UHFFFAOYSA-N | |
| 1201546 | |
| 2-sulfanylidene-1,3-thiazolidin-4-one |
| C3H3NOS2 | |
| MFCD00005488 | |
| rhodanine, 2-thioxothiazolidin-4-one, 2-thioxo-4-thiazolidinone, rhodaninic acid, 4-thiazolidinone, 2-thioxo, 2-thioxo-1,3-thiazolidin-4-one, rhodanic acid, rhodanin, rodanin, 4-oxo-2-thioxothiazoline | |
| CHEBI:8830 | |
| C1C(=O)NC(=S)S1 |
Safety and Handling
GHS H Statement
H318-H302
Causes serious eye damage.
Harmful if swallowed.
P264b-P270-P280i-P301+P312-P305+P351+P338-P330-P501c
H302-H319
EINECSNumber : 205-505-1
RTECSNumber : VI7700000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only