missing translation for 'onlineSavingsMsg'
Learn More

Rhodanine, 98+%

Catalog No. AAA1829314
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1829314 25 g
AAA1829322 100 g
AAA1829336 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1829314 Supplier Thermo Scientific Chemicals Supplier No. A1829314
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 141-84-4 | C3H3NOS2 | 133.183 g/mol

Rhodanine inhibits the multiplication of echovirus 12 and the development of virus-induced morphologic changes. It has been used in tannase assay in cultures of tannin degrading fungi. It possess anticonvulsant, antibacterial, antiviral and antidiabetic activities.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Rhodanine inhibits the multiplication of echovirus 12 and the development of virus-induced morphologic changes. It has been used in tannase assay in cultures of tannin degrading fungi. It possess anticonvulsant, antibacterial, antiviral and antidiabetic activities.

Solubility
Soluble in water (2.25 g/L at 20°C).

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 141-84-4
Molecular Formula C3H3NOS2
Molecular Weight (g/mol) 133.183
MDL Number MFCD00005488
InChI Key KIWUVOGUEXMXSV-UHFFFAOYSA-N
Synonym rhodanine, 2-thioxothiazolidin-4-one, 2-thioxo-4-thiazolidinone, rhodaninic acid, 4-thiazolidinone, 2-thioxo, 2-thioxo-1,3-thiazolidin-4-one, rhodanic acid, rhodanin, rodanin, 4-oxo-2-thioxothiazoline
PubChem CID 1201546
ChEBI CHEBI:8830
IUPAC Name 2-sulfanylidene-1,3-thiazolidin-4-one
SMILES C1C(=O)NC(=S)S1

Specifications

Melting Point 166°C to 171°C
Density 0.868
Odor Odorless
Quantity 25 g
Beilstein 110701
Merck Index 14,8184
Solubility Information Soluble in water (2.25g/L at 20°C).
Formula Weight 133.19
Percent Purity ≥98%
Chemical Name or Material Rhodanine
Hazard Category H302-H319
Hazard Statement GHS H Statement
H318-H302
Causes serious eye damage.
Harmful if swallowed.
Precautionary Statement P264b-P270-P280i-P301+P312-P305+P351+P338-P330-P501c
EINECSNumber 205-505-1
RTECSNumber VI7700000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only