missing translation for 'onlineSavingsMsg'
Learn More

Rhodamine B 95.0+%, TCI America™

Catalog No. R004025G
Change view
Click to view available options
Quantity:
25 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
R004025G 25 g
R0040250G 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. R004025G Supplier TCI America Supplier No. R004025G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 81-88-9
Molecular Formula C28H31ClN2O3
Molecular Weight (g/mol) 479.02
MDL Number MFCD00011931
InChI Key PYWVYCXTNDRMGF-UHFFFAOYSA-N
Synonym rhodamine b, basic violet 10, brilliant pink b, tetraethylrhodamine, rhodamine o, rheonine b, rhodamine fb, calcozine red bx, symulex magenta f, rhodamine
PubChem CID 6694
ChEBI CHEBI:52334
IUPAC Name 9-(2-carboxyphenyl)-3,6-bis(diethylamino)-10λ⁴-xanthen-10-ylium chloride
SMILES [Cl-].CCN(CC)C1=CC2=[O+]C3=CC(=CC=C3C(C3=CC=CC=C3C(O)=O)=C2C=C1)N(CC)CC

Specifications

Melting Point 211°C
Color Green-Yellow
Quantity 25 g
Formula Weight 479.02
Percent Purity ≥95.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Rhodamine B
EINECSNumber (5)-1973&(5)-4056
RTECSNumber BP3675000
TSCA Yes