missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Rhodamine B

Catalog No. AAA1357230
Change view
Click to view available options
Quantity:
50 g
250 g
1000 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1357230 250 g
AAA1357218 50 g
AAA135720B 1000 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1357230 Supplier Thermo Scientific Chemicals Supplier No. A1357230
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 81-88-9 | C28H31ClN2O3 | 479.02 g/mol

Suitable for fluorescent labellingRhodamine B is used as a laser dye, metal chelating agent and an additive to drugs and cosmetics. It also serves as a staining fluorescent dye, which is used as a colorant in inks, paper and plastics. Further, it is used to make pink fluorescent pigments. It is also employed as a biomarker in oral rabies vaccines for wildlife such as raccoons. In addition to this, it is a useful fluorochrome for histology, fluorescence resonance energy transfer (FRET) and mitochondrial probe.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Suitable for fluorescent labellingRhodamine B is used as a laser dye, metal chelating agent and an additive to drugs and cosmetics. It also serves as a staining fluorescent dye, which is used as a colorant in inks, paper and plastics. Further, it is used to make pink fluorescent pigments. It is also employed as a biomarker in oral rabies vaccines for wildlife such as raccoons. In addition to this, it is a useful fluorochrome for histology, fluorescence resonance energy transfer (FRET) and mitochondrial probe.

Solubility
Soluble in water, benzene and ethanol. Slightly soluble in acetone, hydrochloric acid, sodium hydroxide, methyl cellosolve and alcohol.

Notes
Incompatible with strong oxidizing agents.

Chemical Identifiers

CAS 81-88-9
Molecular Formula C28H31ClN2O3
Molecular Weight (g/mol) 479.02
MDL Number MFCD00011931
InChI Key PYWVYCXTNDRMGF-UHFFFAOYSA-N
Synonym Basic Violet 10; C.I. 45170
PubChem CID 6694
ChEBI CHEBI:52334
IUPAC Name [9-(2-carboxyphenyl)-6-(diethylamino)xanthen-3-ylidene]-diethylazanium;chloride
SMILES [Cl-].CCN(CC)C1=CC2=[O+]C3=CC(=CC=C3C(C3=CC=CC=C3C(O)=O)=C2C=C1)N(CC)CC

Specifications

Melting Point ∼210°C (decomposition)
Density 1.31
Beilstein 4119648
Merck Index 14,8183
Solubility Information Soluble in water,benzene and ethanol. Slightly soluble in acetone,hydrochloric acid,sodium hydroxide,methyl cellosolve and alcohol.
Quantity 250 g
Formula Weight 479.02
Chemical Name or Material Rhodamine B
Hazard Category H302-H318
Hazard Statement GHS H Statement
H318-H302
Causes serious eye damage.
Harmful if swallowed.
Precautionary Statement P264b-P270-P280-P301+P312-P305+P351+P338-P310-P330-P501c
EINECSNumber 201-383-9
RTECSNumber BP3675000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only