missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Rhodamine 6G perchlorate, 99%, pure, laser grade

Supplier:  Thermo Scientific™ 419030025

Catalog No. AC419030025

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Explore available promotions

Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Oxidation reagent
Specifications

Specifications

Rhodamine 6G perchlorate
13161-28-9
C28H31ClN2O7
547nm (in methanol)
Authentic
ethyl 2-[3-(ethylamino)-6-ethylimino-2,7-dimethylxanthen-9-yl]benzoate;perchloric acid
13651880
543.01
Pure
Glass bottle
Complies
2.5g
laser grade, 99%
99%
MFCD00013117
HDAFVOZRAUFNQH-UHFFFAOYSA-N
CCNC1=C(C=C2C(=C1)OC3=CC(=NCC)C(=CC3=C2C4=CC=CC=C4C(=O)OCC)C)C.OCl(=O)(=O)=O
543.013
528nm (in methanol)
≥98.5%
104000L/(mole.cm) (lambda max.)
(for use as a laser dye) passes
Pink-Red
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

EINECSNumber : 236-099-4

TSCA : TSCA

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only