missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Rhodamine 123, +99%

Chemical Identifiers

CAS 62669-70-9
Molecular Formula C21H19ClN2O4
Molecular Weight (g/mol) 398.843
MDL Number MFCD00012664
InChI Key SEXIURXGNASJRB-UHFFFAOYSA-N
PubChem CID 16211830
IUPAC Name methyl 2-(3-amino-6-iminoxanthen-9-yl)benzoate;hydrate;hydrochloride
SMILES COC(=O)C1=CC=CC=C1C2=C3C=CC(=N)C=C3OC4=C2C=CC(=C4)N.O.Cl
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC419091000
SDS View Documents
Thermo Scientific™
419091000
100mg Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC419095000
SDS View Documents
Thermo Scientific™
419095000
500mg Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

62669-70-9
398.843
SEXIURXGNASJRB-UHFFFAOYSA-N
methyl 2-(3-amino-6-iminoxanthen-9-yl)benzoate;hydrate;hydrochloride
C21H19ClN2O4
MFCD00012664
16211830
COC(=O)C1=CC=CC=C1C2=C3C=CC(=N)C=C3OC4=C2C=CC(=C4)N.O.Cl

Specifications

62669-70-9
C21H19ClN2O4
15, 8306
Authentic
methyl 2-(3-amino-6-iminoxanthen-9-yl)benzoate;hydrate;hydrochloride
16211830
Extra Pure
99.0% min.
100mg
99+%
MFCD00012664
SEXIURXGNASJRB-UHFFFAOYSA-N
COC(=O)C1=CC=CC=C1C2=C3C=CC(=N)C=C3OC4=C2C=CC(=C4)N.O.Cl
398.843
380.82
Glass bottle
Brown to Brown-Red or Green
Rhodamine 123, 99+%
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Harmful if swallowed.

GHS P Statement:
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

EINECSNumber : 263-687-8

RTECSNumber : ZE0883000

TSCA : TSCA

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only