missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Rhodamine 110, 98+%, pure, laser grade
Supplier: Thermo Scientific™ 419070025
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
| Rhodamine 110 | |
| 98+% | |
| 520nm (in methanol) | |
| Authentic | |
| 2-(3-amino-6-iminoxanthen-9-yl)benzoic acid;hydrochloride | |
| 11617572 | |
| 366.79 | |
| Pure | |
| Glass bottle | |
| White to Yellow |
| 13558-31-1 | |
| C20H15ClN2O3 | |
| JNGRENQDBKMCCR-UHFFFAOYSA-N | |
| C1=CC=C(C(=C1)C2=C3C=CC(=N)C=C3OC4=C2C=CC(=C4)N)C(=O)O.Cl | |
| 366.801 | |
| 498 to 504nm (c=10mg/l in methanol) | |
| ≥98% | |
| 77000 l/(mol.cm) (at lambda max.) min. | |
| (for use as a laser dye) passes | |
| 2.5g |
RUO â Research Use Only