Learn More
Cell Biolabs Inc RhoA/Rac1/Cdc42 Activation Assay Combo Kit, 30 assays (10 per target)
Catalog No. NC0373752
Encompass
Cell Biolabs RhoA/Rac1/Cdc42 Activation Assay Combo Kit provides a simple and fast tool to monitor the activation of RhoA, Rac1 and Cdc42. The kit utilizes easily identifiable Agarose beads (pink in color) to selectively isolate and pull-down the active form of Rho, Rac, or Cdc42 from purified samples or endogenous lysates. Subsequently, the precipitated GTP-form is detected by western blot analysis using an Anti-RhoA , Anti-Rac1, or Anti-Cdc42 specific antibody. Each kit includes an Immunoblot Positive Control and provides sufficient quantities to perform 10 assays of each GTPase target.
Catalog No. NC0373752 Supplier Cell Biolabs Inc Supplier No. STA405
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Cell Biolabs RhoA/Rac1/Cdc42 Activation Assay Combo Kit provides a simple and fast tool to monitor the activation of RhoA, Rac1 and Cdc42. The kit utilizes easily identifiable Agarose beads (pink in color) to selectively isolate and pull-down the active form of Rho, Rac, or Cdc42 from purified samples or endogenous lysates. Subsequently, the precipitated GTP-form is detected by western blot analysis using an Anti-RhoA , Anti-Rac1, or Anti-Cdc42 specific antibody. Each kit includes an Immunoblot Positive Control and provides sufficient quantities to perform 10 assays of each GTPase target.