missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Rho Activation Assay Kit

Catalog No. 17294
Change view
Click to view available options
Quantity:
30 Assays/Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
17294 30 Assays/Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 17294 Supplier MilliporeSigma™ Supplier No. 17294

Includes

  • 100X GTPγS, 10mM
  • 100X GDP, 100mM
  • Mg2+ Lysis/Wash Buffer, 5X
  • Anti-Rho (-A, -B, -C), clone 55
  • Rho Assay Reagent (Rhotekin RBD, agarose)

PP1/PP2A Toolbox for selective in vitro dephosphorylation of proteins

A GST-tagged fusion protein, corresponding to residues 7-89 of mouse Rhotekin Rho Binding Domain, expressed in E. coli. Provided bound to glutathione-agarose. Purity >90%

Specifications

Detection Method Fluorescence, Chemiluminescence, Colorimetric
For Use With (Application) Affinity Binding Assay
Quantity 30 Assays/Kit