Learn More
Cell Biolabs Inc RhoA Activation Assay Kit, 20 assays
Catalog No. NC0258005
Encompass
Cell Biolabs RhoA Activation Assay Kit provides a simple and fast tool to monitor the activation of RhoA. The kit utilizes easily identifiable Rhotekin RBD Agarose beads (pink in color) to selectively isolate and pull-down the active form of Rho from purified samples or endogenous lysates. Subsequently, the precipitated GTP-Rho is detected by western blot analysis using an Anti-RhoA specific antibody. Each kit includes an Immunoblot Positive Control and provides sufficient quantities to perform 20 assays.
Catalog No. NC0258005 Supplier Cell Biolabs Inc Supplier No. STA403A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Cell Biolabs RhoA Activation Assay Kit provides a simple and fast tool to monitor the activation of RhoA. The kit utilizes easily identifiable Rhotekin RBD Agarose beads (pink in color) to selectively isolate and pull-down the active form of Rho from purified samples or endogenous lysates. Subsequently, the precipitated GTP-Rho is detected by western blot analysis using an Anti-RhoA specific antibody. Each kit includes an Immunoblot Positive Control and provides sufficient quantities to perform 20 assays.