Learn More
Innovatek Rheumatoid Factor (RF) Latex Test Kits and Controls
Innovatek Rheumatoid Factor (RF) Latex Test Kits and Controls provide results within 3 minutes
Supplier:  Innovatek 419110521
 View more versions of this product
Catalog No. 35997
Description
Description
  • Fast 2 to 3 minute rotation
  • Complete kit includes all reagents and controls
  • Disposable test cards
  • 6 to 12 months of shelf life
  • Sensitive and specific
Specifications
Chemical Identifiers
Rheumatoid Factor (RF)
≈3 min.
Specifications
Rheumatoid Factor (RF)
1 Vial
≈3 min.
Positive Control
1 mL
Product Suggestions
Product Suggestions