missing translation for 'onlineSavingsMsg'
Learn More

Rhenium wire, 0.5mm (0.02in) dia, 99.97% (metals basis)

Catalog No. AA10312BQ
Change view
Click to view available options
Quantity:
5 cm
25 cm
100 cm
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA10312BQ 5 cm
AA10312BU 25 cm
AA10312BY 100 cm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA10312BQ Supplier Thermo Scientific Chemicals Supplier No. 010312BQ

Form: Approx.4.13g/m | CAS: 7440-15-5 | Re | 186.21 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7440-15-5
Molecular Formula Re
Molecular Weight (g/mol) 186.21
MDL Number MFCD00011195
InChI Key WUAPFZMCVAUBPE-UHFFFAOYSA-N
Synonym rhenium, elementar, renio, unii-7yhu292iny, 7yhu292iny, foil, ion re7, rhenium, ion re7, atom, hydride, pellets
PubChem CID 23947
ChEBI CHEBI:49882
IUPAC Name rhenium
SMILES [Re]

Specifications

Color Silver to Gray
Appearance Silver gray wire
Quantity 5 cm
Odor Odorless
Form Wire
Assay Percent Range 99.97% (Metals basis)
Chemical Name or Material Rhenium
EINECSNumber 231-124-5
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only