missing translation for 'onlineSavingsMsg'
Learn More

Rhenium pentacarbonyl chloride, 98%

Catalog No. AC316310010
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC316310010 1 g Glass bottle
AC316310050 5 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC316310010 Supplier Thermo Scientific Chemicals Supplier No. 316310010

CAS: 14099-01-5 | C5ClO5Re | 361.71 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 14099-01-5
Molecular Formula C5ClO5Re
Molecular Weight (g/mol) 361.71
MDL Number MFCD00013296
InChI Key JQUUAHKBIXPQAP-UHFFFAOYSA-M
Synonym pentacarbonylchlororhenium, carbon monoxide; chlororhenium, pentakis carbon monoxide ; chlororhenium, 5co.clre, rhenium i pentacarbonyl chloride, pentacarbonylchlororhenium i
PubChem CID 6096982
IUPAC Name carbon monoxide;chlororhenium
SMILES [C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].Cl[Re]

Specifications

Color White
Elemental Analysis 97.5% min.
Assay Percent Range 98%
Packaging Glass bottle
Quantity 1 g
Solubility Information Solubility in water: insoluble
Formula Weight 361.71
Percent Purity 98%
Physical Form Fine Powder
Chemical Name or Material Rhenium pentacarbonyl chloride, 98%
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Harmful if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 237-948-1

RUO – Research Use Only