missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Rhenium(IV) sulfide, 99%
CAS: 12038-63-0 | ReS2 | 250.33 g/mol
$744.96 - $3339.96
Chemical Identifiers
| CAS | 12038-63-0 |
|---|---|
| Molecular Formula | ReS2 |
| Molecular Weight (g/mol) | 250.33 |
| MDL Number | MFCD00016307 |
| InChI Key | USWJSZNKYVUTIE-UHFFFAOYSA-N |
| Synonym | rhenium sulfide, rhenium disulphide, disulfanylidenerhenium, dithioxorhenium, rhenium iv sulfide, bis sulfanylidene rhenium |
| PubChem CID | 82864 |
| IUPAC Name | bis(sulfanylidene)rhenium |
| SMILES | S=[Re]=S |
Description
Rhenium(IV) sulfide is used as a diamagnetic semiconductor.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 12038-63-0 | |
| 250.33 | |
| USWJSZNKYVUTIE-UHFFFAOYSA-N | |
| 82864 | |
| S=[Re]=S |
| ReS2 | |
| MFCD00016307 | |
| rhenium sulfide, rhenium disulphide, disulfanylidenerhenium, dithioxorhenium, rhenium iv sulfide, bis sulfanylidene rhenium | |
| bis(sulfanylidene)rhenium |
Specifications
| 12038-63-0 | |
| 2 g | |
| MFCD00016307 | |
| Insoluble in water. Soluble in 1N sodium hydroxide solution. | |
| S=[Re]=S | |
| 250.33 | |
| 250.33 | |
| Odorless | |
| Rhenium (IV) sulfide |
| Powder | |
| ReS2 | |
| rhenium sulfide, rhenium disulphide, disulfanylidenerhenium, dithioxorhenium, rhenium iv sulfide, bis sulfanylidene rhenium | |
| USWJSZNKYVUTIE-UHFFFAOYSA-N | |
| bis(sulfanylidene)rhenium | |
| 82864 | |
| 99% | |
| 7.506 g/mL |
Safety and Handling
EINECSNumber : 234-878-3
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only