missing translation for 'onlineSavingsMsg'
Learn More

Rhenium(III) chloride

Catalog No. p-4782098
Change view
Click to view available options
Quantity:
0.25 g
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA1323001 0.25 g
AA1323003 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA1323001 Supplier Thermo Scientific Chemicals Supplier No. 01323001

CAS: 13569-63-6 | Cl3Re | 292.557 g/mol

It is used as a starting material for synthesis of rhenium complexes. ReCl3 can react with peralkylated benzene-metal complexes that act as electron reservoirs.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 13569-63-6
Molecular Formula Cl3Re
Molecular Weight (g/mol) 292.557
MDL Number MFCD00011196
InChI Key LOIHSHVELSAXQN-UHFFFAOYSA-K
Synonym rhenium iii chloride, rhenium trichloride, rhenium chloride recl3, rhenium 3+ trichloride, recl3, acmc-20ajss
PubChem CID 83581
IUPAC Name trichlororhenium
SMILES Cl[Re](Cl)Cl

Specifications

Color Red
Physical Form Powder
Quantity 0.25 g
Solubility Information Soluble in water.
Formula Weight 292.56
Odor Odorless
Sensitivity Moisture sensitive
Appearance Dark red powder
Chemical Name or Material Rhenium(III) chloride
Hazard Category H315-H319-H335
Hazard Statement In case of contact with eyes, rinse immediately with plenty of water and seek medical advice. Wear suitable gloves.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 236-987-1
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only