missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Rhenium AA Standard, 1000 ppm Re in H2O, Ricca Chemical
Supplier: Ricca Chemical Company ARE1KW100
Specifications
| Rhenium AA Standard | |
| 13598-65-7,7732-18-5 | |
| 100.0,0.15 | |
| MFCD00012377 | |
| [NH4+].O=[Re-](=O)(=O)=O | |
| 268.24 | |
| 1mL = 1mg Re (1, 000ppm Re)NH4ReO4 in H2O |
| Natural Poly Bottle | |
| 98.0,0.14 | |
| H4NO4Re | |
| NLESJKWOAILDGM-UHFFFAOYSA-O | |
| ammonium tetraoxorheniumuide | |
| 3084163 |
Safety and Handling
ShelfLife : 24 months