Learn More
Thermo Scientific™ RheFlex™ Two-Piece PEEK Fittings
Catalog No. 361023904
Encompass
Change view
Click to view available options
Type:
Fitting Set, Short
Catalog No. Type
361023904 Fitting Set, Short
1 options
Catalog No. 361023904 Supplier Thermo Scientific™ Supplier No. 6000055
Thermo Scientific™ RheFlex™ Two-Piece PEEK Fittings provide inert, metal-free connections.
Thermo Scientific™ RheFlex™ Two-Piece PEEK Fittings provide inert, metal-free connections.
  • Slotted back-side of the ferrule is squeezed down onto the tube by the mating conical surface of the nut
  • May be used on 1/16 in. metal or plastic tubing reliably up to 5000 psi
  • Reusable ferrule and nut
Specifications
Material PEEK
For Use With (Equipment) PEEK Tubing
Description 1/16 in. Short fitting set; 5/Pk.
Type Fitting Set, Short
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.