Learn More
Thermo Scientific™ RheFlex™ High-Pressure Fittings
Catalog No. 03251813
Change view
Click to view available options
Type:
Extra Long High Pressure Fittings Set
High Pressure 1/16 in. Ferrule
Quantity:
10/Pk.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Type Quantity
03251813 High Pressure 1/16 in. Ferrule 10/Pk.
03170326 Extra Long High Pressure Fittings Set 10/Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 03251813 Supplier Thermo Scientific™ Supplier No. CMD6000210
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Ensure reliable high-pressure connections with Thermo Scientific™ RheFlex™ High-Pressure Fittings.
Ensure reliable high-pressure connections by using Thermo Scientific™ RheFlex™ High-Pressure Fittings. These fittings are precision machined from 316 stainless steel to be burr- and contaminant-free.
Specifications
Material Stainless Steel
Type High Pressure 1/16 in. Ferrule
For Use With (Equipment) LC Columns
Description Ferrule
Quantity 10/Pk.
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.