missing translation for 'onlineSavingsMsg'
Learn More

National Safety Apparel™ Enespro™ ArcGuard™ RevoLite™ 40 cal Gloves

Catalog No. 17001696
Encompass
Change view
Click to view available options
Size:
Universal
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
17001696 Universal
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 17001696 Supplier National Safety Apparel™ Supplier No. G51NPQH14

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Gloves are constructed from 11.2 oz. RevoLite™ fabric, and have an arc flash rating of 40 cal/cm2.

  • Constructed from 11.2 oz. RevoLite™ fabric
  • Arc rating: 40 cal/cm2

Specifications

Certifications/Compliance NFPA 70E, ASTM F1506, OSHA 1910.269, CSA Z462
Applications Industrial
Color Green
Hand Specific
Material RevoLite
Product Type Arc Flash Gloves
Size Universal
For Use With (Application) Utilities, Manufacturing, Electrical Contractors
Glove Cuff Style Open
Length (English) 14 in.