Learn More
Gilson Reversible Tray PIPETMAN™ M96
Catalog No. FH07003G
Change view
Click to view available options
Includes:
Multi Tray
Rotating Tray
Single Tray
Positions:
384-384
96-384
96-96
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Includes Positions
FH07003G Single Tray 96-384
FH07002G Rotating Tray 96-96
FH07001G Rotating Tray 384-384
FH07005G Rotating Tray 96-384
FH07004G Multi Tray 96-384
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. FH07003G Supplier Gilson Supplier No. FH07003
Only null left
Reversible tray is a convenient tool for stacking, simplifies pipetting procedures and microplate handling needs
  • The single-tray is a convenient tool for stacking and pipetting in microplates, featuring two sides - one for 96-well microplates and the other for filling a 384-well plate.
  • The reversible rotating tray with 96-96, 384-384 well sides is a convenient accessory for various microplate handling needs.
  • The tray is reversible, suitable for 96-well microplates, and can be fitted on either the left or right side of the PIPETMAN™ M96.
  • The rotating tray with reversible 96 left and 384 right side is a convenient accessory for various microplate handling needs.
  • The tray sides can be flipped to fit either 96- or 384-well microplates and can be positioned on the left or right side of the PIPETMAN™ M96.
  • The PIPETMAN™ M96 multi-tray is a convenient accessory that simplifies pipetting procedures, sliding the tray from one microplate to the next.
  • It is compatible with SBS-standard labware, including 96- and 394-well microplates and reservoirs, and is designed to make tasks such as serial dilutions and plate-to-plate transfers easier.
  • Its sliding mechanism enables efficient movement between positions, optimizing space and saving valuable time during pipetting applications.
  • For increased productivity and minimal plastic waste, the use of PIPETMAN™ DIAMOND Tips Blister Refills and Reload Packs is recommended.
Specifications
Product Type Tip Holder
Color Aluminium
Includes Single Tray
Positions 96-384
Quantity 1 Ea.
Weight (English) 0.2 lb.
Weight (Metric) 0.09 kg