Learn More
MilliporeSigma™ Reverse Osmosis Cartridge
Lab Water purification system portfolio offers a broad range of pure and ultrapure water purification systems and services.
Supplier:  MilliporeSigma™ CDRC050HG
Catalog No. CDRC050HG
Only null left
Explore available promotions
Description
Description
  • Designed for scientists working in pharmaceutical, clinical, academic, industrial, research, and government laboratories
  • Can be used in both validated and non-validated environments
  • The Reverse Osmosis membrane removes 95 to 99% of contaminants including ions, particulates, bacteria, and organic molecules
Specifications
Specifications
Water Purification Consumables
Elix™ 35/70/100 systems and RiOs™ 50/100/150/200
Product Suggestions
Product Suggestions