missing translation for 'onlineSavingsMsg'
Learn More

BUCHI FlashPure ID Silica Flash Chromatography Cartridges

Catalog No. 05404800
Change view
Click to view available options
Bed Weight (Metric):
4 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Bed Weight (Metric)
05404800 4 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 05404800 Supplier BUCHI Supplier No. 145146130
Only null left

Includes

20 Pieces

FlashPure ID cartridges are designed for BUCHI Pure chromatography systems. Integrated RFID technology automatically transfers cartridge settings and data to the flash system.

  • High resolution of separation due to high-quality silica
  • Increased fraction purity due to a better separation
  • Faster evaporation of smaller fraction volumes
  • Reproducible performance from batch to batch
  • Increased sample throughput due to high sample loading capacity
  • Solvent saving: required purification yield is achieved faster

Specifications

Material Silica
Type Reveleris
Includes 20 Pc.
Bed Weight (Metric) 4 g
For Use With (Application) Useful for Conventional Applications; Polar Compounds
Max. Pressure 200 psig