Learn More
BUCHI Reveleris™ Empty Solid Loader Tube/Frit
Buchi Reveleris™ Empty Solid Loader Tube/Frit is designed for loading a stationary phase and sample to this cartridge for dry loading chromatography applications
$823.69 - $1,176.74
Specifications
Material Polypropylene
Type Empty Solid Loader
View More Specs
Products 3
Catalog Number Mfr. No. Weight (Metric) Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Weight (Metric) Quantity Price Quantity & Availability  
05402911
View Documents Promotion Details
BUCHI
145142050
3 g 100/Pk.
Pack of 100 Each for $1,176.74
Only null left
05402912
View Documents Promotion Details
BUCHI
145142051
15 g 100/Pk.
Pack of 100 Each for $848.96
Only null left
05402913
View Documents Promotion Details
BUCHI
145147808
40 g 50/Pk.
Pack of 50 Each for $823.69
Only null left
Description
Description
  • Loads your stationary phase and sample to this cartridge for dry loading chromatography applications
  • Use supplied frits to hold sample in place
  • Use the proper sized insertion rod (sold separately) to insert frits
Specifications
Specifications
Polypropylene
Empty Solid Loader