missing translation for 'onlineSavingsMsg'
Learn More

Wells Lamont™ Reusable Full-Finger Nylon Glove Liners

Catalog No. p-7230584
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19086521 Small
19086520 Medium
19086522 Large
19002852 X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 19086521 Supplier Wells Lamont™ Supplier No. M005S.WLC
Only null left

Reusable Full-Finger Nylon Glove Liners are constructed of 100% nylon for a high level of mobility and comfort. Capable of withstanding up to 30 wash cycles without shrinkage.

  • 100% continuous nylon filament construction is thin and comfortable
  • Fully launderable up to 30 times without shrinkage
  • Reduced replacement costs and waste disposal
  • Ideal for use in critical environments and clean rooms
  • Full finger length
  • Ambidextrous
  • Made in the USA

Specifications

Coating Material Uncoated
Color White
Disposable Reusable
Hand Ambidextrous
Latex Free Yes
Material Nylon
Product Type Glove Liners
Size Small
Finger Style Full
For Use With (Application) Critical Environments, Cleanrooms, Electronics Assembly, and Inspection
Glove Cuff Style Knit
Hem Color Green
Series M005.WLC
Show More Show Less