missing translation for 'onlineSavingsMsg'
Learn More
Learn More
- Home
- Products
- Cell Culture Utensils
- Inoculating Loops and Needles
- Thermo Scientific™ Reusable Calibrated Inoculating Loops
Thermo Scientific™ Reusable Calibrated Inoculating Loops
Catalog No. r503740
Change view
Click to view available options
Product Type:
Inoculating Loop
Inoculating Loop Calibration Gauge
Inoculating Needle
Inoculating TB Spade
Insulated Brass Holder
Teasing Needle
Handle Type:
Blue Insulated Handle
Brass Loop Holder
Green Insulated Handle
Not Applicable
Orange Insulated Handle
Purple Insulated Handle
White Insulated Handle
Without Handle
Yellow Insulated Handle
Size:
1/100 mL
1/1000 mL
0.001 mL
2 mm
3 mm
5 mm
6 in.
8 in.
Quantity:
25/Pk.
5/Pk.
Each
16 product options available for selection
| Catalog No. | Product Type | Handle Type | Size | Quantity |
|---|---|---|---|---|
| R503740 | Inoculating Loop | Orange Insulated Handle | 1/100 mL | Each |
| R503060 | Inoculating Needle | Purple Insulated Handle | - | Each |
| R503152 | Inoculating Loop | White Insulated Handle | 2 mm | 5/Pk. |
| R503154 | Inoculating Loop | Green Insulated Handle | 3 mm | Each |
| R503620 | Inoculating Loop | Without Handle | 1/1000 mL | Each |
| R501966 | Insulated Brass Holder | Brass Loop Holder | 6 in. | Each |
| R501968 | Insulated Brass Holder | Brass Loop Holder | 8 in. | Each |
| R503070 | Teasing Needle | Yellow Insulated Handle | - | Each |
| R503080 | Teasing Needle | Orange Insulated Handle | - | Each |
| R503090 | Inoculating TB Spade | Blue Insulated Handle | - | Each |
| R503153 | Inoculating Loop | Green Insulated Handle | 3 mm | Each |
| R503513 | Inoculating Loop | Without Handle | 3 mm | 25/Pk. |
| R503515 | Inoculating Loop | Without Handle | 5 mm | 25/Pk. |
| R503630 | Inoculating Loop | Without Handle | 1/100 mL | Each |
| R503730 | Inoculating Loop | Blue Insulated Handle | 1/1000 mL | Each |
| R507020 | Inoculating Loop Calibration Gauge | Not Applicable | 0.001 mL | Each |
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 16 options available.
Description
Select, inoculate or transfer microbiological sample on media with Thermo Scientific™ Remel™ Reusable Calibrated Inoculating Loops. The gentle, malleable loops offer precise control when selecting or streaking inoculum, without cutting or tearing agar surfaces. The loops are nontoxic, lightweight and are color-coded for easy-recognition and streamlined specimen handling.
- Platinum, twisted nichrome or plastic
- With or without insulated handle
- Some are color coded
Specifications
Specifications
| Volume | 0.01 μL |
| Material | Plastic, Platinum, Nichrome |
| Size | 1/100 mL |
| Color | Orange |
| Product Type | Inoculating Loop |
| Quantity | Each |
| Handle Type | Orange Insulated Handle |