Learn More
Thermo Scientific™ Reusable Calibrated Inoculating Loops
Select, inoculate or transfer a well-defined microbiological sample volume on media plates with these reusable calibrated inoculating loops.
Supplier:  Thermo Scientific™ R503620
 View more versions of this product
Catalog No. R503620
Only null left
Description
Description
Select, inoculate or transfer microbiological sample on media with Thermo Scientific™ Remel™ Reusable Calibrated Inoculating Loops. The gentle, malleable loops offer precise control when selecting or streaking inoculum, without cutting or tearing agar surfaces. The loops are nontoxic, lightweight and are color-coded for easy-recognition and streamlined specimen handling.
  • Platinum, twisted nichrome or plastic
  • With or without insulated handle
  • Some are color coded
Specifications
Specifications
Plastic, Platinum, Nichrome
1/1000 mL
Each
0.001 μL
Inoculating Loop
Without Handle
Product Suggestions
Product Suggestions