Learn More
Thermo Scientific™ Reusable Calibrated Inoculating Loops
Select, inoculate or transfer a well-defined microbiological sample volume on media plates with these reusable calibrated inoculating loops.
Supplier: Thermo Scientific™ RE503152
Description
Select, inoculate or transfer microbiological sample on media with Thermo Scientific™ Remel™ Reusable Calibrated Inoculating Loops. The gentle, malleable loops offer precise control when selecting or streaking inoculum, without cutting or tearing agar surfaces. The loops are nontoxic, lightweight and are color-coded for easy-recognition and streamlined specimen handling.
- Twisted nichrome made
- With or without insulated handle
- Available in different color codes
Specifications
| Nichrome | |
| White | |
| 5/Pk. |
| 2 mm | |
| Inoculating Loop | |
| White Insulated Handle |