Learn More
Thermo Scientific™ Reusable Calibrated Inoculating Loops
Select, inoculate or transfer a well-defined microbiological sample volume on media plates with these reusable calibrated inoculating loops.
Supplier: Thermo Scientific™ RE503730
Description
Select, inoculate or transfer microbiological sample on media with Thermo Scientific™ Remel™ Reusable Calibrated Inoculating Loops. The gentle, malleable loops offer precise control when selecting or streaking inoculum, without cutting or tearing agar surfaces. The loops are nontoxic, lightweight and are color-coded for easy-recognition and streamlined specimen handling.
- Platinum, twisted nichrome or plastic
- With or without insulated handle
- Some are color coded
Specifications
| 0.001 μL | |
| 1/1000 mL | |
| Inoculating Loop | |
| Blue Insulated Handle |
| Plastic, Platinum, Nichrome | |
| Blue | |
| Each |