Learn More
Thermo Scientific™ Reusable Calibrated Inoculating Loops
Select, inoculate or transfer a well-defined microbiological sample volume on media plates with these reusable calibrated inoculating loops.
Supplier:  Thermo Scientific™ RE503153
 Afficher plus de versions de ce produit
Numéro de catalogue. R503153
Il en reste null
Description
Description
Select, inoculate or transfer microbiological sample on media with Thermo Scientific™ Remel™ Reusable Calibrated Inoculating Loops. The gentle, malleable loops offer precise control when selecting or streaking inoculum, without cutting or tearing agar surfaces. The loops are nontoxic, lightweight and are color-coded for easy-recognition and streamlined specimen handling.
  • Twisted nichrome made
  • With or without insulated handle
  • Available in different color codes
Spécifications
Spécifications
Nichrome
Green
Each
3 mm
Inoculating Loop
Green Insulated Handle
missing translation for 'recommendations'
missing translation for 'recommendations'
Videos
FDS