Learn More
Thermo Scientific™ Reusable Calibrated Inoculating Loops
Catalog No. R503060
Change view
Click to view available options
Product Type:
Inoculating Loop
Inoculating Loop Calibration Gauge
Inoculating Needle
Inoculating TB Spade
Insulated Brass Holder
Teasing Needle
Handle Type:
Blue Insulated Handle
Brass Loop Holder
Green Insulated Handle
Not Applicable
Orange Insulated Handle
Purple Insulated Handle
White Insulated Handle
Without Handle
Yellow Insulated Handle
Quantity:
25/Pk.
5/Pk.
Each
Size:
1/100 mL
1/1000 mL
0.001 mL
2 mm
3 mm
5 mm
6 in.
8 in.
16 product options available for selection
Product selection table with 16 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type Handle Type Quantity Size
R503060 Inoculating Needle Purple Insulated Handle Each -
R503152 Inoculating Loop White Insulated Handle 5/Pk. 2 mm
R503154 Inoculating Loop Green Insulated Handle Each 3 mm
R503620 Inoculating Loop Without Handle Each 1/1000 mL
R501966 Insulated Brass Holder Brass Loop Holder Each 6 in.
R501968 Insulated Brass Holder Brass Loop Holder Each 8 in.
R503070 Teasing Needle Yellow Insulated Handle Each -
R503080 Teasing Needle Orange Insulated Handle Each -
R503090 Inoculating TB Spade Blue Insulated Handle Each -
R503153 Inoculating Loop Green Insulated Handle Each 3 mm
R503513 Inoculating Loop Without Handle 25/Pk. 3 mm
R503515 Inoculating Loop Without Handle 25/Pk. 5 mm
R503630 Inoculating Loop Without Handle Each 1/100 mL
R503730 Inoculating Loop Blue Insulated Handle Each 1/1000 mL
R503740 Inoculating Loop Orange Insulated Handle Each 1/100 mL
R507020 Inoculating Loop Calibration Gauge Not Applicable Each 0.001 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 16 options available.
16 options
Catalog No. R503060 Supplier Thermo Scientific™ Supplier No. RE503060
Select, inoculate or transfer a well-defined microbiological sample volume on media plates with these reusable calibrated inoculating loops.
Select, inoculate or transfer microbiological sample on media with Thermo Scientific™ Remel™ Reusable Calibrated Inoculating Loops. The gentle, malleable loops offer precise control when selecting or streaking inoculum, without cutting or tearing agar surfaces. The loops are nontoxic, lightweight and are color-coded for easy-recognition and streamlined specimen handling.
  • Straight
  • Insulated handle
Specifications
Volume No Loop
Material Nichrome
Color Purple
Product Type Inoculating Needle
Quantity Each
Handle Type Purple Insulated Handle