Learn More
Thermo Scientific™ Reusable Calibrated Inoculating Loops
Select, inoculate or transfer a well-defined microbiological sample volume on media plates with these reusable calibrated inoculating loops.
Supplier: Thermo Scientific™ R501968
Description
Select, inoculate or transfer microbiological sample on media with Thermo Scientific™ Remel™ Reusable Calibrated Inoculating Loops. The gentle, malleable loops offer precise control when selecting or streaking inoculum, without cutting or tearing agar surfaces. The loops are nontoxic, lightweight and are color-coded for easy-recognition and streamlined specimen handling.
- Solid brass
Specifications
| No Loop | |
| 8 in. | |
| Insulated Brass Holder | |
| Each |
| Brass | |
| Brass and Black | |
| For use with loops w/o handle | |
| Brass Loop Holder |