Learn More
Thermo Scientific™ Reusable Calibrated Inoculating Loops
Select, inoculate or transfer a well-defined microbiological sample volume on media plates with these reusable calibrated inoculating loops.
Supplier:  Thermo Scientific™ RE501966
 View more versions of this product
Catalog No. R501966
Only null left
Description
Description
Select, inoculate or transfer microbiological sample on media with Thermo Scientific™ Remel™ Reusable Calibrated Inoculating Loops. The gentle, malleable loops offer precise control when selecting or streaking inoculum, without cutting or tearing agar surfaces. The loops are nontoxic, lightweight and are color-coded for easy-recognition and streamlined specimen handling.
  • Solid brass
Specifications
Specifications
No Loop
6 in.
Insulated Brass Holder
Each
Brass
Brass and Black
For use with loops w/o handle
Brass Loop Holder
Product Suggestions
Product Suggestions