missing translation for 'onlineSavingsMsg'
Learn More

CritiCore Protective Wear Reusable Barrier Lab Coats

Catalog No. p-7202968
Change view
Click to view available options
Size:
Medium
6X-Large
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
18111288 Medium
18111295 6X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 18111288 Supplier CritiCore Protective Wear Supplier No. 12L16403BARI01MWHT
Only null left

Specialized lab coats provide optimal fluid resistance. High-density construction protects from permeability.

  • 100% polyester fluid-resistant front and sleeves
  • Quick-release snap front closure
  • Two front pockets and one left breast pocket
  • Side seam openings for pants pocket access
  • Splash Resistant Polyester front and sleeves
  • Breathable PolyCotton back panel for ventilation
  • Industrial launderable
  • Can be used as part of an OSHA Compliance Plan

Specifications

Product Type General Purpose Lab Coat
Size Medium
Gender Unisex
Material Polycotton
Color White
Closure Type Snaps
Cuff Style Knit
Disposable Reusable
Sterility Non-sterile
Resistance Fluids
Style Full-length
No. of Pockets 3
Description Fluid-Resistant Barrier Front, Poly 65 Cotton 35 Back
Collar Type Mandarin
Latex Free Yes
For Use With (Application) Healthcare, Dental, Laboratory, Medical, Veterinary
Show More Show Less